Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units: Alumina Notched Plate
Catalog No. 45001678 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1/Pk.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-678 1/Pk.
45-001-679 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-678 Supplier Cytiva Supplier No. 80613624
Add to Cart
Add to Cart
Specifications
For Use With (Equipment) SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
Type Notched Alumina Plate
Material Alumina
Quantity 1/Pk.
Length (Metric) 10 cm
Width (Metric) 8 cm