Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units:Glass Plate
Catalog No. 45001677 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
SE 250 Mini Vertical Gel Electrophoresis Unit
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
45-001-677 SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
45-001-680 SE 250 Mini Vertical Gel Electrophoresis Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-677 Supplier Cytiva Supplier No. 80613605
Add to Cart
Add to Cart
Specifications
For Use With (Equipment) SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
Type Glass Plate
Material Glass
Shape Rectangular
Length (Metric) 10 cm
Width (Metric) 8 cm