Learn More
Thermo Scientific™ Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets
GSA_VA
Optimize configuration flexibility of 1300 Series A2 Class II Biological Safety Cabinets with these optional stands, UV-lighting, arm rests, foot rests, valves and piping.
$201.00 - $1840.03
Specifications
Certifications/Compliance NSF/ANSI 49, Class II Type A2
Height (English) 30 in. to 37 in.
For Use With (Equipment) Bio Safety Cabinet
View More Specs
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Products 19
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
15-474-224
View Documents
Thermo Scientific™
191127
Adjustable footrest
Each for $645.50
Only null left
Add to Cart
15-474-202
GSA_VA
View Documents
Thermo Scientific™
1911302
Manual Adjustable Stand for 4 ft. Cabinet
Each for $1,247.00
Only null left
Add to Cart
15-474-203
GSA_VA
View Documents
Thermo Scientific™
1911303
Manual Adjustable Stand for 6 ft. Cabinet
Each for $1,512.38
Only null left
Add to Cart
15-474-204
View Documents
Thermo Scientific™
1911304
Combustible Gas Valve for Universal Piping Option
Each for $280.00
Only null left
Add to Cart
12-223-024
View Documents
Thermo Scientific™
1911305
Noncombustible Gas Valve for Universal Piping Option
Each for $201.00
Only null left
Add to Cart
15-474-206
GSA_VA
missing translation for 'viewDocuments'
Thermo Scientific™
1911306
Vacuum Valve for Universal Piping Option
Each for $274.00
Only null left
Add to Cart
15-474-208
GSA_VA
missing translation for 'viewDocuments'
Thermo Scientific™
1911308
Combustible Gas Valve for Standard Applications
Each for $301.50
Only null left
Add to Cart
15-474-209
GSA_VA
missing translation for 'viewDocuments'
Thermo Scientific™
1911309
Noncombustible Gas Valve for Standard Applications
Each for $296.50
Only null left
Add to Cart
15-474-210
GSA_VA
missing translation for 'viewDocuments'
Thermo Scientific™
1911310
Vacuum Valve for Standard Applications
Each for $301.50
Only null left
Edge
Add to Cart
15-474-211
missing translation for 'viewDocuments'
Thermo Scientific™
1911311
Water Valve for Standard Applications
Each for $293.50
Only null left
Add to Cart
15-474-212
missing translation for 'viewDocuments'
Thermo Scientific™
1911312
IV bag holder kit with 12 hooks (for 4 ft., 5 ft., and 6 ft. cabinets)
Each for $664.00
Only null left
Add to Cart
15-474-213
GSA_VA
missing translation for 'viewDocuments'
Thermo Scientific™
1911313
Arm Rest (Set of 2)
Each for $279.00
Only null left
Add to Cart
15-474-216
GSA_VA
missing translation for 'viewDocuments'
Thermo Scientific™
1911316
Thimble duct exhaust transition for 3 ft., 4 ft., and 5 ft. cabinets
Each for $1,243.00
Only null left
Add to Cart
15-474-217
GSA_VA
missing translation for 'viewDocuments'
Thermo Scientific™
1911317
Thimble Duct Exhaust Transition for 6 ft. Cabinet
Each for $1,412.20
Only null left
Add to Cart
15-474-218
missing translation for 'viewDocuments'
Thermo Scientific™
1911318
UV Light Option, Qi
Each for $645.50
Only null left
Add to Cart
15-474-219
missing translation for 'viewDocuments'
Thermo Scientific™
1911319
UV light option for 3 ft. and 4 ft. Cabinet
Each for $641.50
Only null left
Add to Cart
15-474-325
missing translation for 'viewDocuments'
Thermo Scientific™
1911322
Rolling Caster Stand for 4 ft. Cabinet
Each for $1,631.00
Only null left
Add to Cart
15-474-326
missing translation for 'viewDocuments'
Thermo Scientific™
1911346
Rolling Caster Stand for 6 ft. Cabinet
Each for $1,840.03
Only null left
Add to Cart
15-474-327
missing translation for 'viewDocuments'
Thermo Scientific™
1911368
UV Light for 6 ft. Cabinet
Each for $531.50
Only null left
Add to Cart
Description
Description
Utilize these Thermo Scientific™ Accessories for Series 1300 Class II, Type A2 Biological Safety Cabinets to increase the flexibility of said cabinets. Accessories include optional stands, UV-lighting, arm rests foot rests, valves and piping.
Stands
  • Manual and electric stands provide comfortable work surface height of 30-37in. (75-95cm) for seated or standing applications
  • Manual stands are adjustable in 1in. increments; electric stands are infinitely adjustable
  • Rolling caster stand provides comfortable fixed worksurface height of 34in. (86cm) for seated applications; designed for limited cabinet mobility for cleaning purposes
  • Stands are included with packages
UV Light
  • Provides safe and easy method for cabinet disinfection
  • Factory-installed; order with cabinet
  • Included with packages
Armrest and Footrest
  • Stainless-steel armrests (set of two) provide ergonomic forearm support and extended reach inside cabinet with less airflow disruption; included with packages
  • Footrest provides ergonomic foot and posture support during seated use
IV Bag Holder Kit
  • Bar and 12 hooks
  • For hanging IV bags near interior ceiling of cabinet
Valves
  • For standard applications or universal piping
  • Standard-application valves have long stem pipe fitting appropriate for side-access media services
  • Short piping stem on universal-piping valves will not penetrate cabinet side wall
  • Combustible gas valve: Rated for combustible gas dispensing inside cabinet chamber
  • Noncombustible gas valve: Rated for noncombustible gas dispensing inside cabinet chamber
  • Vacuum valve: Rated for routing vacuum inside cabinet chamber
  • Water valve: Rated for water dispensing inside cabinet chamber
Thimble Exhaust Duct Transition
  • Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
  • Method recommended by NSF/ANSI™ 49 for externally exhausting Class II, Type A2 biological safety cabinets
Universal Piping for Exhaust Systems
  • Allows cabinet to be connected to an external exhaust for removal of volatile toxic chemicals or radionuclides used in cabinet
  • Factory-installed; must be ordered with cabinet
Universal Piping for Air/Gas Service
  • Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
  • Two pre-plumbed service ports per side only
  • Factory-installed; must be ordered with cabinet
  • For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Specifications
Specifications
NSF/ANSI 49, Class II Type A2
30 in. to 37 in.
Bio Safety Cabinet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.