Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Corporation AccessQuick RT-PCR System

Catalog No. PRA1703 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Sample Type: RNA samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRA1703 Sample Type: RNA samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA1703 Supplier Promega Corporation Supplier No. A1703
Only null left
Add to Cart
Add to Cart

Convenient RT-PCR in one tube contains Tfl DNA Polymerase, dNTPs, magnesium sulfate, and reaction buffer. Combine with AMV RT, RNA template, primers, and water for efficient RT-PCR, including separate AMV RT for no-RT control reactions.

  • Convenient RT-PCR system in a single tube.
  • Contains all necessary components for efficient RT-PCR.
  • Includes Tfl DNA Polymerase, dNTPs, magnesium sulfate, and reaction buffer.
  • Separate AMV RT provided for no-RT control reactions.
  • Simple and quick setup for RNA template analysis.
  • Ensures high sensitivity and specificity.
  • Compatible with various RNA samples.
  • Streamlined workflow reduces hands-on time.
  • Produces consistent and reliable results.
  • Ideal for gene expression studies and other applications.

Specifications

Concentration 2X (Master Mix)
Detection Method qPCR, RT-PCR
Form Liquid
Format One-tube RT-PCR system
PCR Method 1-step RT-PCR
Polymerase Tfl DNA Ploymerase
Reaction Speed Fast or Standard
Technique RT-PCR
Kit Type Master mix for RT-PCR
No. of Tests 500 Tests
Product Type RT-PCR system
Quantity 12.5 mL
Species Various (dependant on sample)
Sufficient For 500 RT-PCR reactions
For Use With (Application) RT-PCR
Compatibility Wide range of RNA, mRNA, total RNA etc.
Fidelity (vs. Taq) Tfl DNA Polymerase
Final Product Type cDNA for subsequent PCR amplification
No. of Reactions 500 Reactions
Purification Target RNA to cDNA
Reaction Format Single tube
Reagent Type Tfl DNA Polymerase, dNTPs, magnesium sulfate, reaction buffer, and AMV reverse transcriptase.
Sample Type RNA samples
Starting Material RNA
Volume 50 μL per reaction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.