Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega AccessQuick™ RT-PCR System

Catalog No. PRA1702 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
20 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRA1702 100 Reactions
PR-A17-01 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRA1702 Supplier Promega Supplier No. A1702
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

AccessQuick Master Mix, 2X; AMV reverse transcriptase; Nuclease-free water

The AccessQuick RT-PCR System is an easy and convenient master mix for one-tube RT-PCR.

  • Convenient RT-PCR in One Tube
  • Mix contains Tfl DNA Polymerase, dNTPs, magnesium sulfate and reaction buffer
  • Separate AMV RT provided for important no-RT control reactions
  • Simply combine AccessQuick™ Master Mix with AMV RT, RNA template, primers and water for efficient RT-PCR

For laboratory use.

Specifications

Concentration 2X (Master Mix)
Amplicon Size 323
Content And Storage • 2X Master Mix, 3 x 1 mL
• Reverse Transcriptase, 1 x 500 μL
• Nuclease-free Water, 2 x 1.25 mL

Store at –30° to –10°C.
Detection Method qPCR, RT-PCR
Form Liquid
Format Kit
PCR Method 1-step RT-PCR
Polymerase Tfl DNA Polymerase
Technique RT-PCR
DNase DNase-free
Host Cell E. coli
Includes Label or Dye No
Kit Type AccessQuick™ RT-PCR system
Label or Dye Unlabeled
No. of Tests 100 Tests
Optimal Reaction Temperature 68°C to 72°C
Primer Gene-Specific Primers
Product Line AccessQuick
Product Type RT-PCR Kit
Reverse Transcriptase AMV Reverse Transcriptase
Ribonuclease H Activity Yes
Sufficient For 100 Reactions
For Use With (Application) RT-PCR
Activation Time 2 min.
Compatibility RT-PCR
Fidelity (vs. Taq) 1X
Final Product Type cDNA
Hot Start No
No. of Reactions 100 Reactions
Overhang 3'-A
Reaction Format Separate Components
Reagent Type RT-PCR Reagent
Sample Type RNA
Starting Material RNA
Volume 50 μL
Source Tfl DNA Polymerase and AMV Reverse Transcriptase
Storage Buffer AMV/Tfl 5X Reaction Buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.