Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Acclaim™ PepMap™ 100 C18 HPLC Columns

Catalog No. 164944
Change view
Click to view available options
Length (Metric):
5 mm
20 mm
50 mm
150 mm
250 mm
500 mm
750 mm
Diameter (Metric):
0.05 mm
0.075 mm
0.1 mm
0.2 mm
0.3 mm
75 μm
1 mm
Particle Size:
2 μm
3 μm
5 μm
Product Type:
Analytical Column
Analytical Column, NanoViper
Analytical nano Column
Column
Trap Cartridge
Trap Column
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size Product Type
16-494-4 250 mm 0.05 mm 2 μm Column
DX164568 150 mm 0.075 mm 3 μm Analytical Column
DX164569 250 mm 0.075 mm 3 μm Analytical Column
DX164570 500 mm 0.075 mm 3 μm Analytical Column
DX164572 150 mm 1 mm 3 μm Analytical Column
DX160434 5 mm 1 mm 5 μm Trap Cartridge
DX164213 150 mm 0.2 mm 5 μm Trap Column
DX164199 150 mm 0.1 mm 5 μm Trap Column
DX164537 150 mm 0.3 mm 2 μm Analytical Column
DX164534 150 mm 0.075 mm 2 μm Analytical Column
DX164560 50 mm 0.3 mm 2 μm Analytical Column
DX164535 150 mm 0.075 mm 3 μm Trap Column
16-494-1 250 mm 0.075 mm 2 μm Analytical Column
16-494-2 500 mm 0.075 mm 2 μm Analytical Column
03-050-825 50 mm 1 mm 2 μm Analytical Column, NanoViper
16-494-6 20 mm 75 μm 3 μm Trap Column
16-494-3 150 mm 0.05 mm 2 μm Analytical Column
DX164564 150 mm 0.1 mm 5 μm Trap Column
16-494-0 150 mm 0.075 mm 2 μm Analytical Column
03-050-828 150 mm 1 mm 2 μm Analytical Column
16-494-5 500 mm 0.05 mm 2 μm Column
DX164939 750 mm 0.075 mm 2 μm Analytical nano Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 16-494-4 Supplier Thermo Scientific™ Supplier No. 164944
Add to Cart
Add to Cart

Use for high-resolution tryptic, natural, and synthetic peptide analyses. Its high loading capacity is exceptional for low abundant peptides in complex proteomic samples.

Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ PepMap™ 100 C18 LC Columns. PepMap LC columns offer lengths targeting fast or high resolution peptide separation.

  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows highest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
  • Preassembled with nanoViper column inlet fittings for easy column installation

Key advantages of nanoViper fittings

  • Ease of use, no sleeve for small capillaries
  • Tool-free assembly
  • No column damage from overtightening: capable of withstanding pressures up to 1000 bars and is compatible with third-party valves and unions
  • No experimental failure due to bad connections

Ideal for high-resolution peptide mapping of tryptic, natural, and synthetic peptides as well as LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology.

  • High loading capacity
  • Use for the analysis of low abundant peptides often found in complex proteomics samples
  • Select the column length and I.D. for your throughput, sensitivity, and amount of sample available requirements
  • Choose from 50, 150, 250, 500mm lengths and 50, 75, 300 and 1000μm I.D.
  • Superior protein and peptide separations
  • Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity
  • Outstanding separation efficiency
  • Superior resolution and recovery
  • Easy installation with nanoViper™ Fingertight Fittings

PepMap nanocolumns are also available pre-assembled with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fitting for a dead-volume-free connection to 1000 bar.

  • Requires no tools
  • Provides virtually zero-dead-volume connections
  • Withstands pressures up to a 1000 bar

About Nano, capillary, and micro Acclaim PepMap columns

Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and microcolumns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary, and microcolumn in a variety of chemistries for peptides and protein analysis.

Use PepMap Trap columns for desalting and preconcentration of peptides before LC separation with MS detection. The trap columns provide the highest efficiencies for one-dimensional peptide mapping experiments and 2D-LC analyses.

TRUSTED_SUSTAINABILITY

Specifications

Column Format NanoViper Column
Diameter (Metric) 0.05 mm
For Use With LC-MS, ESI-MS
Length (Metric) 250 mm
Max. Pressure 17400 psi (1200 bar)
Particle Size 2 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 450 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line nanoLC
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
For Use With (Application) Nano-flow Peptide Mapping
Product Type Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.