Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Acclaim™ 300 C18 HPLC Columns
GREENER_CHOICE

Catalog No. 03254089
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-254-089 150 mm 3 mm
03-050-806 50 mm 2.1 mm
DX060264 150 mm 2.1 mm
03-050-807 50 mm 4.6 mm
DX060266 150 mm 4.6 mm
03-254-090 10 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-254-089 Supplier Thermo Scientific™ Supplier No. 063684
Add to Cart
Add to Cart

Use these wide-pore C18 columns to achieve high-resolution protein separations.

Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim™ 300 C18 LC Columns. The 3μm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5μm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.

High-Resolution Separation of Proteins and Peptides
  • High-performance C18 bonding chemistry on 300Å pore silica
  • Application tested for suitability in peptide mapping
  • Reproducible for dependable results
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 .5 and 7.5
About the Acclaim Family of HPLC Columns
  • Complementary selectivity, high column efficiencies and ultrahigh resolution
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases and specialty phases including wide-pore
  • Performance tested to ensure high quality and reliability
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 3 mm
Flow Rate 1 mL/min
For Use With LC/MS
Length (Metric) 150 mm
Max. Pressure 4500 psi (310 bar)
Particle Size 3 μm
pH 2.5 to 7.5
Pore Size 300 Å
Quantity Each
Surface Area 100 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line Acclaim
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 8%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Material Glass Lined Tubing
Technique Bio-LC
Green Features FSC
Product Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.