Learn More
Thermo Scientific™ Acclaim™ 300 C18 HPLC Columns

Description
Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim™ 300 C18 LC Columns. The 3μm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5μm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.
- High-performance C18 bonding chemistry on 300Å pore silica
- Application tested for suitability in peptide mapping
- Reproducible for dependable results
- Extremely low bleed, fully compatible with MS
- Stable between pH 2 .5 and 7.5
- Complementary selectivity, high column efficiencies and ultrahigh resolution
- Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases and specialty phases including wide-pore
- Performance tested to ensure high quality and reliability
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Flow Rate | 1 mL/min |
| For Use With | LC/MS |
| Length (Metric) | 10 mm |
| Max. Pressure | 6000 psi (414 bar) |
| Particle Size | 3 μm |
| pH | 2.5 to 7.5 |
| Pore Size | 300 Å |
| Quantity | Pack of 2 |
| Surface Area | 100 m²/g |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.