Learn More
Thermo Scientific™ Acclaim™ AmG C18 HPLC Columns

Description
Get a high-performance, reversed-phase columns for aminoglycoside analysis. The robust and rugged Thermo Scientific™ Acclaim™ AmG C18 HPLC column provides exceptional tolerance required for aminoglycosides ion-pairing chromatographic separations. These silica-based C18 columns are optimized for various aminoglycoside antibiotic analyses, including drug purity characterization and quantification, therapeutic drug monitoring, and residual control testing in different matrices. These columns are stable at low pH, high temperature, organic solvents, and aqueous mobile phases.
The Acclaim AmG C18 column is specifically designed for ion-pairing reversed-phase HPLC (IP-RPLC) analysis of aminoglycoside antibiotics using volatile perfluorinated carboxylic acids as the ion-pairing reagent, such as 100 mM trifluoroacetic acid (TFA). The Acclaim AmG C18 stationary phase is based on a proprietary C18 bonding technology that provides extremely high stability towards acidic conditions. This specialty column exhibits excellent selectivity and high resolution for the aminoglycoside antibiotics analysis. In addition, it is easy to use because simple aqueous mobile phase (e.g. 100 mM TFA) without organic solvent and pH adjusting buffer is sufficient in most cases.
- Excellent selectivity for aminoglycosides
- Superior tolerance towards acidic conditions
- High efficiency and throughput
- Ease of use
The column can be used with UV, CAD, ELSD detectors
Specifications
Specifications
| Column Format | Guard Column |
| Diameter (Metric) | 3 mm |
| Length (Metric) | 10 mm |
| Max. Pressure | 8000 psi (550 bar) |
| Particle Size | 3 μm |
| pH | 0.5 to 10 |
| Pore Size | 120 Å |
| Quantity | 2 Guard Cartridges/Pk. (Requires Guard Cartridge Holder 069580) |
| Temperature | <80°C |
| USP Type | L1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.