Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCN1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002531 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002531 Supplier Novus Biologicals Supplier No. NB1002531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACCN1 Beta Polyclonal specifically detects ACCN1 Beta in Human, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen ACCN1 Beta
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:300-3:000
Formulation PBS with 0.05% Sodium Azide
Gene Alias Acid-sensing ion channel 2, Amiloride-sensitive brain sodium channel, amiloride-sensitive cation channel 1, neuronal, ASIC2a, ASIC2Mammalian degenerin homolog, BNAC1, BNaC1ACCN, BNC1Amiloride-sensitive cation channel neuronal 1, degenerin, hBNaC1, MDEGBrain sodium channel 1, neuronal amiloride-sensitive cation channel 1
Gene Symbols ASIC2
Host Species Rabbit
Immunogen CSGPGRFMAREQ (amino acid residues 15-26).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 40
Test Specificity This is specific to the ASIC 2B protein between amino acid residues 15-26.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.