Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCN2 Antibody (S271-44), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP222409IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222409IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222409IR Supplier Novus Biologicals Supplier No. NBP222409IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACCN2 Monoclonal specifically detects ACCN2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ACCN2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S271-44
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Acid-sensing ion channel 1, acid-sensing ion channel 1a protein, amiloride-sensitive cation channel 2, neuronal, ASIC, ASIC1ASIC1A, BNAC2, BNaC2ACCN2 variant 3, Brain sodium channel 2, Cation channel, amiloride-sensitive, neuronal, 2, hBNaC2
Gene Symbols ASIC1
Host Species Mouse
Immunogen Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 41
Test Specificity Detects approx 60kDa.
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.