Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Accucore™ 150-C4 HPLC Columns

Catalog No. 03898128
Change view
Click to view available options
Length (Metric):
10 mm
30 mm
50 mm
100 mm
150 mm
Diameter (Metric):
0.075 mm
2.1 mm
3 mm
4.6 mm
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-898-128 150 mm 4.6 mm
03-898-117 30 mm 2.1 mm
03-898-120 50 mm 2.1 mm
03-898-124 100 mm 3 mm
03-898-125 100 mm 4.6 mm
03-898-126 150 mm 2.1 mm
03-898-132 150 mm 0.075 mm
03-898-129 10 mm 2.1 mm
03-898-130 10 mm 3 mm
03-898-119 30 mm 4.6 mm
03-898-122 50 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 03-898-128 Supplier Thermo Scientific™ Supplier No. 16526154630
Add to Cart
Add to Cart

Use this specially designed solid core column with a C4 functionality for biomolecules that bind too tightly to C8 or C18 columns.

Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.

  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Significantly lower hydrophobic retention than C18
  • Ideal for optimal retention of proteins and larger peptides

Ordering Information:

Other custom column dimensions are available. Please call your local Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
For Use With HPLC
Length (Metric) 150 mm
Max. Pressure 14500 psi (1000 bar)
Particle Shape Solid Core
Particle Size 2.6 μm
pH 2 to 9
Pore Size 150 Å
Quantity Each
Surface Area 80 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Product Line Accucore
Packing Material Ultrapure Silica
Carbon Load 2%
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.