Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AccuPrime™ SuperMix II

Catalog No. p-4926959
Encompass_Preferred
Click to view available options
Quantity:
200 Reactions

Requires

Template and primers

AccuPrime SuperMix II is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.

AccuPrime SuperMix II is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half. For accurate PCR performance, AccuPrime SuperMix II sets the standard. You'll amplify your DNA targets cycle after cycle, eliminating troubleshooting and rework.

In addition to a powerful DNA polymerase, AccuPrime SuperMix I includes Platinumanti-DNA polymerase antibodies that inhibit polymerase activity, providing an automatic hot-start, and a thermal stable accessory protein that enhances specific primer-template hybridization during every cycle of PCR. This unique combination allows for success, even under previously suboptimal conditions.

AccuPrime SuperMix II is designed for amplification of genomic DNA amplicons, plasmid DNA, or cDNA templates up to 4 kb in length. Alternatively, AccuPrime SuperMix I is optimized to amplify a tighter range of targets less than 200 bp in length.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 Reactions
Includes AccuPrime SuperMix II is supplied at 2X concentration to allow approximately 50% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 200 amplification reactions of 25 μL each are provided. AccuPrime SuperMix II Components: 40 mM Tris-HCl (pH 8.4); 100 mM KCl; 3 mM MgCl2, 400 μM each dGTP, dATP, dTTP, dCTP; AccuPrime Taq DNA Polymerase; thermostable AccuPrime protein; stabilizers.

200 Reactions: 2 x 1.25 mL.
Store at -20°C in a non-frost-free freezer.
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.