Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ACD |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15315420
![]() |
Novus Biologicals
NBP15315420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP153154
![]() |
Novus Biologicals
NBP153154 |
100 μL |
Each for $487.50
|
|
|||||
Description
ACD Polyclonal specifically detects ACD in Human samples. It is validated for Western Blot.Specifications
ACD | |
Polyclonal | |
Rabbit | |
Q96AP0 | |
65057 | |
Synthetic peptides corresponding to ACD(adrenocortical dysplasia homolog (mouse)) The peptide sequence was selected from the middle region of ACD (NP_001075955). Peptide sequence KNRPPFPRTGATRGAQEPCSVWEPPKRHRDGSAFQYEYEPPCTSLCARVQ. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
adrenocortical dysplasia homolog (mouse), Pip1, PIP1Tint1, POT1 and TIN2 organizing protein, POT1 and TIN2-interacting protein, Ptop, PTOPTpp1, TIN2 interacting protein 1, TINT1adrenocortical dysplasia protein homolog, TPP1 | |
ACD | |
IgG | |
58 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title