Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE-2 Antibody (1034734), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB305476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305476 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305476 Supplier Novus Biologicals Supplier No. FAB10822MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for ELISA, Flow Cytometry.

Specifications

Antigen ACE-2
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 1034734
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived human ACE-2, Gln18-Ser740, Accession # Q9BYF1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 59272
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.