Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE-2 Antibody (2818I), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB296486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296486 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296486 Supplier Novus Biologicals Supplier No. FAB34372L
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ACE-2 Monoclonal antibody specifically detects ACE-2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry

Specifications

Antigen ACE-2
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2818I
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived mouse ACE-2, Gln18-Thr740, Accession # NP_081562
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 59272
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.