Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE-2 Antibody (379131), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB310482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310482 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310482 Supplier Novus Biologicals Supplier No. FAB9334APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ACE-2 Monoclonal antibody specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen ACE-2
Applications Flow Cytometry
Classification Monoclonal
Clone 379131
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Host Species Rat
Immunogen Mouse myeloma cell line, NS0-derived human ACE-2, Gln18-Ser740, Accession # Q9BYF1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 59272
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.