Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE-2 Antibody (OTI4D2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP272117UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272117UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272117UV Supplier Novus Biologicals Supplier No. NBP272117UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ACE-2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI4D2
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Gene Symbols ACE2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 18-237 of human ACE-2 (NP_068576) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 59272.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.