Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE/CD143 Antibody (171417), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345791 Supplier Novus Biologicals Supplier No. FAB929MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen ACE/CD143
Applications CyTOF, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 171417
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunoprecipitation, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ACE/CD143, aa 30-1261
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, CD Markers, Cell Biology, Endocrinology, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1636
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.