Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, 1 M, pH 3.5, PROMO, Thermo Scientific Chemicals

Catalog No. J60340APPM
Click to view available options
Quantity:
500 mL

Applications

Acetate, 1M buffer solution, pH, 3.5 is used in dexoyribionucleic acid (DNA) and ribonucleic acid (RNA) precipitation. It is also used in the preparation of gel stains for protein gel electrophoresis.

Solubility

Soluble in water and ethanol.

Notes

Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1 M
Form Liquid
Quantity 500 mL

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.