Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, 1 M, pH 4.0, PROMO, Thermo Scientific Chemicals

Catalog No. J60104APPM
Click to view available options
Quantity:
500 mL

Applications

Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. Other reported uses. Used in the purification and precipitation of nucleic acids; Used in protein crystallization. Staining of gels in protein gel electrophoresis and HPLC; Mordant dyeing. DNA microarray studies of E. coli response; Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents and as a substrate of acetokinase.

Solubility

Fully miscible with water. Soluble in ethanol.

Notes

Store in cool. Keep container tightly closed in a dry and well-ventilated place.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1 M
pH Range 4
Form Liquid
Quantity 500 mL

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.