Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, 1 M, pH 5.5, PROMO, Thermo Scientific Chemicals

Catalog No. J61033APPM
Click to view available options
Quantity:
500mL

Applications

Acetate, 1 M buffer solution, pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

Solubility

Soluble in water (246 g/L) at 20°C, and ethanol.

Notes

Contains sodium acetate and glacial acetic acid, pH 5.5

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1 M
Form Liquid
Quantity 500mL

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.