Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt, 99+%, for molecular biology, anhydrous, DNAse, RNAse and Protease free

Catalog No. p-3734322
Click to view available options
Quantity:
250 g
2.5 kg

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Absorbance (0.1 M in water), (0.1 M in water) at 0nm, 0.01 max. at 260nm, 0.01 max. at 280nm
pH 7 to 9.2 (5% soln. at 25°C)
Melting Point 324.0°C
CAS Min % 99.0
Color White
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3CO2Na
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name sodium;acetate
Formula Weight 82.03
Percent Purity ≥99%
Grade Molecular Biology
Quantity 250 g
Flash Point >250°C
Infrared Spectrum Authentic
Loss on Drying 1% max. (120°C)
Packaging Plastic bottle
Physical Form Crystalline Solid
Chemical Name or Material Acetic acid, sodium salt, Anhydrous, DNAse, RNAse and Protease Free
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.