Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoacetyl CoA synthetase Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB323997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323997 Supplier Novus Biologicals Supplier No. NBP299709JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acetoacetyl CoA synthetase Polyclonal antibody specifically detects Acetoacetyl CoA synthetase in Human samples. It is validated for ELISA

Specifications

Antigen Acetoacetyl CoA synthetase
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias acetoacetate-CoA ligase, acetoacetyl-CoA synthetase, ACSF1FLJ41251, Acyl-CoA synthetase family member 1, EC 6.2.1, EC 6.2.1.16, FLJ12389, homolog of C. elegans supressor of ras 5 (sur-5), Protein sur-5 homolog, SUR-5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Acetoacetyl CoA synthetase (Accession#: NP_076417.2; Met1-Phe672).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65985
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.