Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone/Acetonitrile, 80:20 Mixture v/v Made with HPLC Grade Components, Reagents

Catalog No. 01335920
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-920 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-920 Supplier Reagents Holdings Llc Supplier No. CA1200004C
Only null left
Add to Cart
Add to Cart

Acetone/Acetonitrile, 80:20 Mixture v/v Made with HPLC Grade Components, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1 , 75-05-8
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone/Methyl cyanide, 2-Propanone//Methyl cyanide
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Color Colorless
Boiling Point 56°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7879
Grade Reagent
Chemical Name or Material Acetone/Acetonitrile, 80:20 Mixture v/v Made with HPLC Grade Components
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.