Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.1% formic acid, CHROMASOLV™ LC-MS, Solstice

Catalog No. 60048223 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
GL45 Glass Bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-223 4 x 2.5 L GL45 Glass Bottle
60-048-215 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-223 Supplier Solstice Supplier No. 346684X2.5L/DEL

CHROMASOLV™ LC-MS, LabReady™ Solvent Blend

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8 , 64-18-6
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00149894
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Min % >99.0000%
CAS Max % >99%, 0.1000%
Density 0.7767 g/cm3
Quantity 4 x 2.5 L
Flash Point 8°C
Packaging GL45 Glass Bottle
Linear Formula ZnSO4 · 7H2O
UN Number UN1648
Boiling Point 82°C
Percent Purity >99.5%
Grade LC-MS
Chemical Name or Material Acetonitrile with 0.1% formic acid, CHROMASOLV™ LC-MS
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution
EINECSNumber 200-835-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.