Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 44%, Aqueous, HPLC, Reagents

Catalog No. 01335925
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-925 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-925 Supplier Reagents Holdings Llc Supplier No. CA1081004C
Only null left
Add to Cart
Add to Cart

Acetonitrile, 44%, Aqueous, HPLC, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 75-05-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methyl cyanide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 76°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9065
Percent Purity 44%
Grade Reagent
Chemical Name or Material Acetonitrile, 44%, Aqueous, HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.