Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Anhydrous, For Biotechnology, MilliporeSigma™

Catalog No. p-7083017 Shop All MilliporeSigma Products
Click to view available options
Quantity:
4 L
20 L
Packaging:
Plastic Barrel
Plastic Bottle

Recommended Storage

Store at room temperature

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Boiling Point 81.6°C (1013 hPa)
Quantity 4 L
Flash Point 2°C
Packaging Plastic Barrel
Physical Form Clear Liquid, Free From Particulates
Vapor Pressure 97 hPa (20°C)
Water 10ppm
Formula Weight 41.05
Percent Purity 0.999
EINECSNumber 200-835-2
RTECSNumber AL7700000
Recommended Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.