Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetonitrile, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 4 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 2 ppm |
| Titratable Acid | 8 μeq/g |
| Titratable Base | 0.6 μeq/g |
| Water | 0.0002 |
| Percent Purity | 0.999 |
| Grade | High Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction