Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile (HPLC), Fisher Chemical

Catalog No. a998212 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
2.5 L
4 L
19 L
Packaging:
Amber Glass
Amber Glass Bottle
NOWPak™ I
Safe-Cote™ Amber Glass Bottle
Wide Neck Amber Bottle

HPLC mobile phase for analytical and preparative separations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 0.01 max. at 254 nm, 0.02 max. at 220 nm, 0.04 max. at 210 nm, 0.05 max. at 205 nm, 0.07 max. at 200 nm, 0.15 max. at 195 nm, 1 max. at 190 nm, Pass Test
Color Colorless
Decomposition Information >500°C
Boiling Point 81°C
Quantity 2.5 L
Flash Point 12.8°C (55°F)
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation 1.0 ppm max.
Assay Percent Range Assay: 99.9% min by GC
Specific Gravity 0.781
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Linear Formula CH3CN
Vapor Pressure 97 mbar at 20°C
UN Number 1648; Colorless
Viscosity 0.36 cP at 20°C
Water 0.01% max.
Merck Index 15, 69
Solubility Information Soluble in water
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.