Learn More
Acetonitrile (HPLC), Fisher Chemical PROMO
Shop All Fisherbrand ProductsSpecifications
Specifications
| Melting Point | -46°C | 
| Absorbance | 0.01 Max at 254 nm, 0.02 max at 220 nm, 0.04 max at 210 nm, 0.05 max at 205 nm, 0.07 max at 200 nm, 0.15 max at 195 nm, 1 max at 190 nm, Pass Test | 
| Color | Colorless | 
| Decomposition Information | >500°C | 
| Boiling Point | 81°C | 
| Quantity | 4 L | 
| Flash Point | 12.8°C (55°F) | 
| Identification | Pass Test | 
| Packaging | Amber Glass Bottle | 
| Physical Form | Liquid | 
| Show More | 
Safety and Handling
                        
                        
                            
                        
                        | Hazard Statement | DANGER! | 
| Precautionary Statement | Emergency Overview: Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air. NFPA Health:2 Flammability:3 Instability:0  | 
                                
| DOTInformation | DOT Class 3, : Flammable Liquid | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.