Learn More
Acetonitrile, LC-MS Grade, 99.8%
Description
Solvent for LC-MS analysisAcetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -48°C to -45°C |
| Color | Colorless |
| Density | 0.786 g/mL |
| Boiling Point | 80°C to 82°C |
| Quantity | 4 L |
| Flash Point | 5°C (41°F) |
| Odor | Pungent |
| Physical Form | Liquid |
| Refractive Index | 1.344 |
| UN Number | UN1648 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H312+H332-H319 |
| Hazard Statement | GHS H Statement H225-H302-H312-H332-H319 Highly flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ACETONITRILE |
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.