Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetonitrile, Reagent, ACS, 99.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
Steel Can
Description
Spectrum™ Chemical Acetonitrile, Reagent, ACS is a colorless liquid with low viscosity and low chemical reactivity and is used in chemical labs as a polar aprotic solvent in organic synthesis. Acetonitrile is a medium-polarity solvent with a convenient liquid range that is miscible with water. As an ACS grade Reagent, Spectrum Chemical manufactured acetonitrile is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications
Specifications
| CAS Max % | 100% |
| Appearance | Clear |
| Quantity | 20 L |
| Packaging | Steel Can |
| Residue after Evaporation | 0.005% |
| Titratable Acid | 8 μq/g |
| Titratable Base | 0.6 μeq/g |
| UN Number | UN1648 |
| Water | 0.3% |
| Percent Purity | 99.5% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction