Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Ultra Low Water, BakerDRY™, J.T. Baker™

Catalog No. p-5390984 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-191 100 mL Glass Bottle
02-002-192 1 L Narrow Mouth Amber Glass Bottle, w/ Septum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-002-191 Supplier Avantor Supplier No. 903510
Only null left
Add to Cart
Add to Cart

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Quantity 100 mL
Flash Point 2.00°C
Packaging Glass Bottle
Specific Gravity 1L = 0.79kg
Formula Weight 41.05
Grade BakerDRY
Chemical Name or Material Acetonitrile, Ultra Low Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.