Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl β-Catenin (Lys49) Rat anti-Human, Clone: mAb-K49-Ac, MilliporeSigma™

Rat Monoclonal Antibody

$191.34 - $481.79

Specifications

Antigen Acetyl β-Catenin (Lys49)
Clone mAb-K49-Ac
Applications Dot Blot, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE182925
View Documents
MilliporeSigma
MABE182925UG
25 μg
Each for $191.34
Only null left
Add to Cart
 
MABE1829100
View Documents
MilliporeSigma
MABE1829100UG
100 μg
Each for $481.79
Only null left
Add to Cart
 
Description

Description

Acetyl β-Catenin (Lys49) Monoclonal antibody specifically detects Acetyl β-Catenin (Lys49) in Human samples. It is validated for Dot Blot, Immunoprecipitation, Western Blotting.
Specifications

Specifications

Acetyl β-Catenin (Lys49)
Dot Blot, Immunoprecipitation, Western Blot
Unconjugated
Rat
Human
BSA-conjugated linear peptide corresponding to 19 amino acids surrounding acetylated lysine 49 from human -catenin.
Primary
Stable for 1 year at 2-8°C from date of receipt.
mAb-K49-Ac
Monoclonal
Purified
RUO
Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2b κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.