Learn More
Novus Biologicals™ Acetyl-CoA Carboxylase Activity Assay Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Gene ID (Entrez) | 31 |
| Quantity | 96 Tests |
| Synonym | ACAC, Acac1, ACACAD, ACC, ACC1, ACCA, ACC-alpha, ACCalphaACACalpha, acetyl-CoA carboxylase 1, acetyl-Coenzyme A carboxylase alpha, hACC1 |
| Species | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.