Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-CoA Carboxylase alpha/ACACA Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291921 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291921 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291921 Supplier Novus Biologicals Supplier No. NB10055246APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acetyl-CoA Carboxylase alpha/ACACA Polyclonal specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen Acetyl-CoA Carboxylase alpha/ACACA
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate APC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2
Host Species Rabbit
Immunogen Maps to a region between residue 1925 and 1975 of human acetyl-Coenzyme A carboxylase alpha using the numbering given in entry NP_942131.1 (GeneID 31).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 31
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.