Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Lysine Antibody (3P8N3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB166062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB166062 Supplier Novus Biologicals Supplier No. NBP315483
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Acetyl-Lysine Monoclonal antibody specifically detects Acetyl-Lysine in All species samples. It is validated for Western Blot

Specifications

Antigen Acetyl-Lysine
Applications Western Blot
Classification Monoclonal
Clone 3P8N3
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias acetyl Lysine, AC-LYS-OH, Lysine, N-acetyl lysine
Host Species Mouse
Immunogen Acetylated BSA
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Target Species All species
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.