Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Lysine Antibody (7F8), Biotin, Novus Biologicals™
SDP

Catalog No. NB11096876B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096876B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11096876B Supplier Novus Biologicals Supplier No. NB11096876B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Acetyl-Lysine Monoclonal antibody specifically detects Acetyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Acetyl-Lysine
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7F8
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias acetyl Lysine, AC-LYS-OH, Lysine, N-acetyl lysine
Host Species Mouse
Immunogen Acetylated KLH
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Detects proteins containing acetylated lysine residues in ELISA and western blots. Does not detect non-acetylated lysine residues.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.