Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl MLL1 (KMT2A) (Lys 1130/1133) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | Acetyl MLL1 (KMT2A) (Lys 1130/1133) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Dot Blot, Inhibition Assays, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE344-25UG
|
MilliporeSigma
ABE34425UG |
25 μg |
Each for $192.35
|
|
|||||
ABE344-100UG
|
MilliporeSigma
ABE344100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-Acetyl MLL1 (KMT2A) (Lys 1130/1133), Cat. No. ABE344, is a highly specific rabbit polyclonal antibody that targets Histone-lysine N-methyltransferase 2A (MLL1) acetylated on Lysine 1130/1133 and has been tested for use in Dot Blot, Peptide Inhibition Assay, and Western Blotting.Specifications
Acetyl MLL1 (KMT2A) (Lys 1130/1133) | |
Dot Blot, Inhibition Assays, Western Blot | |
Unconjugated | |
Rabbit | |
Epigenetics & Nuclear Function | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.09% sodium azide. | |
NP_005924 | |
KLH-cpnugated linear peptide corresponding to 12 amino acids surrounding acetylated lysines 1130 and 1133 from the N-terminal half of human Histone-lysine N-methyltransferase 2A. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Histone-lysine N-methyltransferase 2A;EC: 2.1.1.43;Lysine N-methyltransferase 2A;ALL-1;CXXC-type zinc finger protein 7;Myeloid/lymphoid or mixed-lineage leukemia;Myeloid/lymphoid or mixed-lineage leukemia protein 1;Trithorax-like protein;Zinc finger protein HRX | |
KMT2A;ALL1;CXXC7;HRX;HTRX;MLL;MLL1;TRX1 | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects human Histone-lysine N-methyltransferase 2A acetylated on lysine 1130 and 1133. | |
431.76 kDa calculated. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title