Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylacetaldehyde dimethyl acetal, 92%

Catalog No. AC292330010
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

CAS: 5436-21-5 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5436-21-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00008789
InChI Key PJCCSZUMZMCWSX-UHFFFAOYSA-N
Synonym 4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal
PubChem CID 228548
IUPAC Name 4,4-dimethoxybutan-2-one
SMILES COC(CC(C)=O)OC

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point -82°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 178°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4140 to 1.416
Linear Formula CH3COCH2CH(OCH3)2
Quantity 1 L
Specific Gravity 0.99
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with most common organic solvents
Viscosity 1.503 mPa.s (20°C)
Formula Weight 132.16
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Acetylacetaldehyde dimethyl acetal, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-605-1
RTECSNumber EL7592500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.