Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylacetaldehyde dimethyl acetal, tech. 90%

Catalog No. AAA120580B
Click to view available options
Quantity:
100 g
250 g
1000 g

CAS: 5436-21-5 | C6H12O3 | 132.16 g/mol

Acetaldehyde dimethyl acetal is used as a flavoring agent. It is also used in the preparation of (R)-4,4-dimethoxy-2-butanol, pyrazoles and pyrimidines. Further, it is used in the preparation of N,N-diethyl-[2-(4-fluorophenyl)-5-methylpyrazolo[1,5-a]-pyrimidin-3-yl]acetamide and acetoacetaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde dimethyl acetal is used as a flavoring agent. It is also used in the preparation of (R)-4,4-dimethoxy-2-butanol, pyrazoles and pyrimidines. Further, it is used in the preparation of N,N-diethyl-[2-(4-fluorophenyl)-5-methylpyrazolo[1,5-a]-pyrimidin-3-yl]acetamide and acetoacetaldehyde.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with acids and strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5436-21-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00008789
InChI Key PJCCSZUMZMCWSX-UHFFFAOYSA-N
Synonym 4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal
PubChem CID 228548
IUPAC Name 4,4-dimethoxybutan-2-one
SMILES COC(CC(C)=O)OC

Specifications

Melting Point -83°C
Density 0.996
Boiling Point 170°C to 172°C
Flash Point 57°C (134°F)
Refractive Index 1.415
Quantity 1000 g
UN Number UN1224
Beilstein 1702372
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 132.16
Percent Purity 90%
Grade Technical
Chemical Name or Material Acetylacetaldehyde dimethyl acetal
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 226-605-1
RTECSNumber EL7592500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.