Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcholinesterase/ACHE Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB141896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB141896 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB141896 Supplier Novus Biologicals Supplier No. NB600911G
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Bovine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Acetylcholinesterase/ACHE
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Host Species Goat
Immunogen Acetylcholinesterase/ACHE [Bovine Erythrocytes] (Uniprot: P23795)
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 43
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.