Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AcetylCoenzymeACarboxylaseBeta Rabbit anti-Human, Clone: ST53-08, Novus Biologicals™
SDP

Catalog No. NBP267545 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267545 Supplier Novus Biologicals Supplier No. NBP267545

Rabbit Monoclonal Antibody

Acetyl Coenzyme A Carboxylase Beta Monoclonal specifically detects Acetyl Coenzyme A Carboxylase Beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AcetylCoenzymeACarboxylaseBeta
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ST53-08
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias ACC2ACC-beta, ACCBacetyl-CoA carboxylase 2, acetyl-CoA carboxylase beta, acetyl-Coenzyme A carboxylase beta, EC 6.4.1.2, HACC275
Gene Symbols ACACB
Host Species Rabbit
Immunogen Synthetic peptide within Human Acetyl Coenzyme A Carboxylase Beta aa 382-427/ 2346. (SwissProt: Q13085 Human; SwissProt: Q5SWU9 Mouse; SwissProt: P11497 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 32
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.