Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ACF |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15731320
![]() |
Novus Biologicals
NBP15731320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157313
![]() |
Novus Biologicals
NBP157313 |
100 μL |
Each for $487.50
|
|
|||||
Description
ACF Polyclonal specifically detects ACF in Human samples. It is validated for Western Blot.Specifications
ACF | |
Polyclonal | |
Rabbit | |
Q9NQ94-2 | |
29974 | |
Synthetic peptides corresponding to A1CF(APOBEC1 complementation factor) The peptide sequence was selected from the N terminal of A1CF. Peptide sequence MESNHKSGDGLSGTQKEAALRALVQRTGYSLVQENGQRKYGGPPPGWDAA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ACF65, ACFASPACF64, apo-B RNA editing protein, APOBEC1 complementation factor, apobec-1 complementation factor (ACF) (ASP), APOBEC-1 stimulating protein, APOBEC1CF, APOBEC1-stimulating protein, MGC163391 | |
A1CF | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title