Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACF1/BAZ1A Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560074
Encompass_Preferred
Change view
Click to view available options
:
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-0074 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0074 Supplier Bethyl Laboratories, Inc Supplier No. A301318AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

BAZ1A may play a role in transcriptional regulation. May be involved in the formation or maintece of heterochromatin playing a critical role in developmental control.

Specifications

Antigen ACF1/BAZ1A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene BAZ1A
Gene Accession No. Q9NRL2
Gene Alias ACF1, ATP-dependent chromatin remodeling protein, ATP-dependent chromatin-remodeling protein, ATP-utilizing chromatin assembly and remodeling factor 1, bromodomain adjacent to zinc finger domain, 1A, CHRAC subunit ACF1, hACF1, hWALp1, WALp1, WCRF180, williams syndrome transcription factor-related chromatin-remodeling factor 180
Gene Symbols BAZ1A
Host Species Rabbit
Immunogen Between 725 and 775
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11177
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.