Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Achaete Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179448
Description
Achaete Polyclonal specifically detects Achaete in Drosophila samples. It is validated for Western Blot.Specifications
Achaete | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
990 E5 F1, Ac, ac achaete, Ac/Sc, ASC, AS-C T5, AS-C T5ac, ascT5, CG3796, Dmel\CG3796, EG:125H10.3, Hw, sc/T5, T5 | |
Rabbit | |
Affinity purified | |
RUO | |
30981 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_476824 | |
ac | |
The specific Immunogen is proprietary information. Peptide sequence FNGPSVIRRNARERNRVKQVNNGFSQLRQHIPAAVIADLSNGRRGIGPGA. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: House fly: 92%; Blue blowfly: 92%; African malaria mosquito: 85%; Red flour beetle: 85%; Silk moth: 85%; Yellowfever mosquito: 85%; Mosquito: 85%; Harpegnathos saltator: 85%; Camponotus floridanus: 85%; Southern house mosquito: 85%; Caribbean spiny lobster: 85%; Mediterranean fruit fly: 84%; Starlet sea anemone: 83%; Drosophila quadrilineata: 78%; Body louse: 78%; Drosophila sordidula: 78%; Drosophila pavani: 78%; Drosophila paramelanica: 78%; Drosophila nigromelanica: 78%; Drosophila micromelanica: 78%; Drosophila macrospina: 78%; Drosophila gaucha: 78%; Drosophila euronotus: 78%; Drosophila colorata: 78%; Triops longicaudatus: 78%. | |
Human, Rat, Drosophila | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction