Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Achaete Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Achaete |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17944820
![]() |
Novus Biologicals
NBP17944820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179448
![]() |
Novus Biologicals
NBP179448 |
100 μL |
Each for $487.50
|
|
|||||
Description
Achaete Polyclonal specifically detects Achaete in Drosophila samples. It is validated for Western Blot.Specifications
Achaete | |
Polyclonal | |
Rabbit | |
NP_476824 | |
30981 | |
The specific Immunogen is proprietary information. Peptide sequence FNGPSVIRRNARERNRVKQVNNGFSQLRQHIPAAVIADLSNGRRGIGPGA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
990 E5 F1, Ac, ac achaete, Ac/Sc, ASC, AS-C T5, AS-C T5ac, ascT5, CG3796, Dmel\CG3796, EG:125H10.3, Hw, sc/T5, T5 | |
ac | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title