Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Acifran
GREENER_CHOICE

Catalog No. 176250
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Hypolipidemic agent; agonist for the GPR109A (HM74A) and GPR109B receptors

Hypolipidemic agent. Exhibits higher potency than nicotinic acid and clofibrate. Full and potent agonist at the human orphan GPCR HM74A/GPR109A and GPR109B (EC50 values are 1.3 and 4.2 μM respectively). Reduces serum triglycerides and circulating LDL-cholesterol in vivo, without affecting liver weight or liver enzymes.

Chemical Identifiers

CAS 72420-38-3
Molecular Formula C12H10O4

Specifications

Quantity 50 mg
Formula Weight 218.21
Purity >99%
Molecular Weight (g/mol) 218.208
Chemical Name or Material Acifran
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.