Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acinus Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500217 0.1 mg
NB018182 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500217 Supplier Novus Biologicals Supplier No. NB500217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot.

Specifications

Antigen Acinus
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 -1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107
Gene Symbols ACIN1
Host Species Rabbit
Immunogen A peptide made to the C-terminal sequence RNRQLEREKRREHS.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 22985
Test Specificity Reacts with the C-terminus of Acinus.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.